Assay Details
| Gene Symbol: Obp2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099301.1 | NP_001092771.1 | 3 - 4 | 334 | 112 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Obp2b| Gene Symbol: Obp2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099301.1 | NP_001092771.1 | 3 - 4 | 334 | 112 | |
Set Membership: |
|