Assay Details
| Gene Symbol: Ssc5d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173008.2 | NP_766596.1 | 10 - 11 | 2368 | 70 | |
| GenBank mRNA | AK079906.1 | - | 10 - 11 | 2369 | 70 | |
| AK165666.1 | - | 10 - 11 | 2430 | 70 | ||
| BC141044.1 | - | 10 - 11 | 2366 | 9055656 | 70 | |
| EU850434.1 | - | 10 - 11 | 2369 | 70 | ||
More Information
Set Membership: |
|

Ssc5d
Genomic Map