Assay Details
| Gene Symbol: Brsk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003920.3 | NP_001003920.2 | 10 - 11 | 1176 | 84 | |
| GenBank mRNA | AY533671.1 | - | 10 - 11 | 1018 | 84 | |
| BC086636.1 | - | 10 - 11 | 1175 | 6402113 | 84 | |
More Information
Updated Mapping Information
Set Membership: |
|

Brsk1
Genomic Map