Assay Details
| Gene Symbol: Brsk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003920.3 | NP_001003920.2 | 12 - 13 | 1440 | 71 | |
| GenBank mRNA | AY533671.1 | - | 12 - 13 | 1282 | 71 | |
| BC086636.1 | - | 12 - 13 | 1439 | 6402113 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Brsk1
Genomic Map