Assay Details
| Gene Symbol: Brsk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003920.3 | NP_001003920.2 | 18 - 19 | 2325 | 75 | |
| GenBank mRNA | AY533671.1 | - | 18 - 19 | 2167 | 75 | |
| BC086636.1 | - | 18 - 19 | 2324 | 6402113 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
G2/M transition of mitotic cell cycle
protein phosphorylation
cellular response to DNA damage stimulus
cell cycle
neurotransmitter secretion
nervous system development
axonogenesis
response to UV
phosphorylation
peptidyl-serine phosphorylation
establishment of cell polarity
neuron differentiation
G2 DNA damage checkpoint
intracellular signal transduction
neuron projection morphogenesis
centrosome duplication
nucleotide binding
magnesium ion binding
protein kinase activity
protein serine/threonine kinase activity
protein binding
ATP binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups
protein kinase binding
gamma-tubulin binding
metal ion binding
tau-protein kinase activity
G2/M transition of mitotic cell cycle
protein phosphorylation
cellular response to DNA damage stimulus
cell cycle
neurotransmitter secretion
nervous system development
axonogenesis
response to UV
phosphorylation
peptidyl-serine phosphorylation
establishment of cell polarity
neuron differentiation
G2 DNA damage checkpoint
intracellular signal transduction
neuron projection morphogenesis
centrosome duplication
nucleotide binding
magnesium ion binding
protein kinase activity
protein serine/threonine kinase activity
protein binding
ATP binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups
protein kinase binding
gamma-tubulin binding
metal ion binding
tau-protein kinase activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2134092 | BC086636.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2134092 | BC086803.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2134092 | NM_001003920.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2051991 | BC086636.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2051991 | BC086803.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2051991 | NM_001003920.3 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 336142 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 950245 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 60244 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 326711 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 385846 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 478707 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 407738 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 467701 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 251544 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 117773 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 655135 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 487379 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 709230 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 409983 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 433933 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 205921 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 167312 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 1614 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 146496 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 175038 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P118082 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P155162 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P133267 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2134092 | BC086636.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2134092 | BC086803.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2134092 | NM_001003920.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2051991 | BC086636.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2051991 | BC086803.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2051991 | NM_001003920.3 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950010 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950014 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950006 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950017 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950009 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950015 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950011 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950008 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950007 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950005 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950019 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950020 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P155162 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06114298 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P133267 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P118082 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P155162 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P118082 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P133267 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06090588 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06090585 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949974 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950104 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950113 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950132 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950164 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950148 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950162 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950015 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950076 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950065 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950000 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950146 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950118 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950041 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950106 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950169 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950131 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950120 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950128 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950056 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950088 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950071 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950096 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949971 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950054 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950155 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950141 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949963 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950047 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950069 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950049 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950158 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950019 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950027 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950024 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950026 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950092 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950028 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949958 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950111 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949993 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950053 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950044 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950160 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950036 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950064 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950134 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949961 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950014 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950121 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950084 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950110 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950094 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950124 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950012 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949982 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949968 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950170 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950029 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950086 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949992 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950107 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950022 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950008 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950078 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950073 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950081 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950152 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949978 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950009 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950089 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949998 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949970 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950150 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949987 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949967 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950060 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949985 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950040 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950034 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949996 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950006 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950161 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950127 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950010 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949964 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949960 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950048 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950137 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950007 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950116 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949955 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24950007 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24949992 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06659473 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06372934 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06240739 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06475022 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06195444 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24950019 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24950035 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06635562 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05908894 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06372941 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06090595 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06127781 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06195443 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06090589 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06544283 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24949996 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24950040 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06014906 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24950037 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24950005 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06451551 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06014913 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06021450 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06372938 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06014909 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06544286 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06014912 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24950026 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06090593 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06014905 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06481212 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24949993 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10549696 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10549694 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10549683 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10549688 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10549690 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10549682 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10549681 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10549693 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10549684 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10549686 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10549689 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10549692 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10549698 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10549699 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10549691 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10549687 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10549697 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10549685 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10549695 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559645 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10549680 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1439892_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10549679 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10559644 | - | - |
| Mouse Genome 430 2.0 Array | 1439892_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1439892_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 381979_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 381979_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700000501.mm.1 | TC0700000038.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700000501.mm.1 | NM_001003920.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700000501.mm.1 | NM_001003920.3 | Refseq |
| Mouse Expression Set 430 | 1439892_PM_at | - | - |
| Mouse Expression Set 430 | 1439892_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1439892_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1439892_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 347514 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2540940 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3104328 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3104328 | - | - |

Brsk1
Genomic Map