Assay Details
| Gene Symbol: Brsk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003920.3 | NP_001003920.2 | 4 - 5 | 609 | 70 | |
| NM_001168572.1 | NP_001162044.1 | 4 - 5 | 609 | 70 | ||
| GenBank mRNA | AY533671.1 | - | 4 - 5 | 451 | 70 | |
| BC086636.1 | - | 4 - 5 | 608 | 6402113 | 70 | |
| EU016556.1 | - | 4 - 5 | 457 | 70 | ||
| EU016557.1 | - | 4 - 5 | 451 | 70 | ||
| EU016558.1 | - | 4 - 5 | 451 | 70 | ||
| EU586326.1 | - | 4 - 5 | 639 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Brsk1
Genomic Map