Assay Details
| Gene Symbol: Ptprh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_207270.2 | NP_997153.2 | 17 - 18 | 2824 | 62 | |
| GenBank mRNA | AB217856.1 | - | 16 - 17 | 2497 | 62 | |
| AK078884.1 | - | 16 - 17 | 2501 | 62 | ||
More Information
Set Membership: |
|

Ptprh
Genomic Map