Assay Details
| Gene Symbol: Dtnbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025772.4 | NP_080048.2 | 2 - 3 | 236 | 91 | |
| GenBank mRNA | AJ404859.1 | - | 2 - 3 | 235 | 91 | |
| AK150306.1 | - | 2 - 3 | 232 | 91 | ||
| AK159656.1 | - | 2 - 3 | 361 | 91 | ||
| BC018350.1 | - | 2 - 3 | 227 | 4195234 | 91 | |
| BC048682.1 | - | 2 - 3 | 281 | 91 | ||
| BC058574.1 | - | 2 - 3 | 209 | 6487720 | 91 | |
More Information
Set Membership: |
|

Dtnbp1
Genomic Map