Assay Details
| Gene Symbol: Dtnbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025772.4 | NP_080048.2 | 8 - 9 | 793 | 71 | |
| GenBank mRNA | AJ404859.1 | - | 8 - 9 | 792 | 71 | |
| AK010924.1 | - | 4 - 5 | 405 | 71 | ||
| AK017400.1 | - | 4 - 5 | 571 | 71 | ||
| AK159656.1 | - | 8 - 9 | 918 | 71 | ||
| AK180780.1 | - | 1 - 2 | 87 | 71 | ||
| AK189627.1 | - | 1 - 2 | 87 | 71 | ||
| AY265461.1 | - | 4 - 5 | 571 | 71 | ||
| BC018350.1 | - | 8 - 9 | 784 | 4195234 | 71 | |
| BC048682.1 | - | 8 - 9 | 838 | 71 | ||
| BC058574.1 | - | 8 - 9 | 766 | 6487720 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dtnbp1
Genomic Map