Assay Details
| Gene Symbol: Igf2bp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023670.3 | NP_076159.3 | 13 - 14 | 2012 | 74 | |
| GenBank mRNA | AB046173.1 | - | 13 - 14 | 1529 | 74 | |
| AK011689.1 | - | 13 - 14 | 1793 | 74 | ||
| AK088465.1 | - | 13 - 14 | 1771 | 74 | ||
| AK147140.1 | - | 13 - 14 | 2016 | 74 | ||
| BC045138.1 | - | 13 - 14 | 1642 | 6306448 | 74 | |
| BC049082.1 | - | 13 - 14 | 1678 | 5706238 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Igf2bp3
Genomic Map