Assay Details
| Gene Symbol: Gpnmb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053110.4 | NP_444340.3 | 2 - 3 | 432 | 72 | |
| GenBank mRNA | AF322054.1 | - | 2 - 3 | 270 | 72 | |
| AJ251685.1 | - | 2 - 3 | 317 | 72 | ||
| AK076347.1 | - | 2 - 3 | 301 | 72 | ||
| AK079220.1 | - | 274 | 72 | |||
| AK149700.1 | - | 2 - 3 | 301 | 72 | ||
| AK149740.1 | - | 2 - 3 | 339 | 72 | ||
| AK150739.1 | - | 2 - 3 | 301 | 72 | ||
| AK154617.1 | - | 2 - 3 | 336 | 72 | ||
| AK159165.1 | - | 2 - 3 | 331 | 72 | ||
| AK159270.1 | - | 2 - 3 | 339 | 72 | ||
| AK159747.1 | - | 2 - 3 | 301 | 72 | ||
| AK159908.1 | - | 2 - 3 | 307 | 72 | ||
| AK170506.1 | - | 2 - 3 | 331 | 72 | ||
| AK171347.1 | - | 2 - 3 | 307 | 72 | ||
| AK171441.1 | - | 2 - 3 | 301 | 72 | ||
| AK171465.1 | - | 2 - 3 | 301 | 72 | ||
| AK171474.1 | - | 2 - 3 | 301 | 72 | ||
| AK171517.1 | - | 2 - 3 | 419 | 72 | ||
| AK171608.1 | - | 2 - 3 | 301 | 72 | ||
| AK171618.1 | - | 2 - 3 | 301 | 72 | ||
| AK190327.1 | - | 1 - 2 | 95 | 72 | ||
| AK202963.1 | - | 1 - 2 | 111 | 72 | ||
| BC026375.1 | - | 2 - 3 | 306 | 4164706 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gpnmb
Genomic Map