Assay Details
| Gene Symbol: Gpnmb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053110.4 | NP_444340.3 | 5 - 6 | 909 | 80 | |
| GenBank mRNA | AF322054.1 | - | 5 - 6 | 747 | 80 | |
| AJ251685.1 | - | 5 - 6 | 794 | 80 | ||
| AK044764.1 | - | 5 - 6 | 803 | 80 | ||
| AK076347.1 | - | 5 - 6 | 778 | 80 | ||
| AK079220.1 | - | 751 | 80 | |||
| AK149700.1 | - | 5 - 6 | 778 | 80 | ||
| AK149740.1 | - | 5 - 6 | 816 | 80 | ||
| AK150739.1 | - | 5 - 6 | 778 | 80 | ||
| AK154617.1 | - | 5 - 6 | 813 | 80 | ||
| AK159165.1 | - | 5 - 6 | 808 | 80 | ||
| AK159270.1 | - | 5 - 6 | 816 | 80 | ||
| AK159747.1 | - | 5 - 6 | 778 | 80 | ||
| AK159908.1 | - | 5 - 6 | 784 | 80 | ||
| AK170506.1 | - | 5 - 6 | 808 | 80 | ||
| AK171347.1 | - | 5 - 6 | 784 | 80 | ||
| AK171441.1 | - | 5 - 6 | 778 | 80 | ||
| AK171465.1 | - | 5 - 6 | 778 | 80 | ||
| AK171474.1 | - | 5 - 6 | 778 | 80 | ||
| AK171517.1 | - | 5 - 6 | 896 | 80 | ||
| AK171608.1 | - | 5 - 6 | 778 | 80 | ||
| AK171618.1 | - | 5 - 6 | 778 | 80 | ||
| AK179971.1 | - | 2 - 3 | 186 | 80 | ||
| AK192937.1 | - | 1 - 2 | 53 | 80 | ||
| AK193818.1 | - | 1 - 2 | 53 | 80 | ||
| AK196346.1 | - | 1 - 2 | 53 | 80 | ||
| AK201982.1 | - | 1 - 2 | 133 | 80 | ||
| AK204190.1 | - | 1 - 2 | 53 | 80 | ||
| AK215880.1 | - | 1 - 2 | 53 | 80 | ||
| BC026375.1 | - | 5 - 6 | 783 | 4164706 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gpnmb
Genomic Map