Assay Details
| Gene Symbol: Tra2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198102.2 | NP_932770.2 | 1 - 2 | 215 | 78 | |
| GenBank mRNA | AK171708.1 | - | 1 - 2 | 197 | 78 | |
| BB873436.1 | - | 1 - 2 | 215 | 78 | ||
| BC057448.1 | - | 1 - 2 | 173 | 4016620 | 78 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the AK171708.1,BC057448.1,NM_198102.2 transcripts.
This assay is in the 5'UTR region of the AK171708.1,BC057448.1,NM_198102.2 transcripts.
Set Membership: |
|

Tra2a
Genomic Map