Assay Details
| Gene Symbol: Tra2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198102.2 | NP_932770.2 | 6 - 7 | 942 | 80 | |
| GenBank mRNA | AK164994.1 | - | 1 - 2 | 83 | 80 | |
| AK171708.1 | - | 5 - 6 | 2162 | 80 | ||
| BC023018.1 | - | 3 - 4 | 322 | 80 | ||
| BC058764.1 | - | 7 - 8 | 1207 | 80 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tra2a
Genomic Map