Assay Details
| Gene Symbol: Malsu1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029353.1 | NP_083629.1 | 2 - 3 | 485 | 76 | |
| GenBank mRNA | AK010366.1 | - | 2 - 3 | 427 | 76 | |
| AK031878.1 | - | 2 - 3 | 436 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Malsu1
Genomic Map