Assay Details
| Gene Symbol: Cdan1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026891.2 | NP_081167.2 | 16 - 17 | 2378 | 64 | |
| GenBank mRNA | AK034144.1 | - | 1 - 2 | 71 | 64 | |
| AK157271.1 | - | 16 - 17 | 2379 | 64 | ||
| BC117555.1 | - | 14 - 15 | 1663 | 40098039 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cdan1
Genomic Map