Assay Details
| Gene Symbol: Gas7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109657.3 | NP_001103127.1 | 4 - 5 | 604 | 63 | |
| NM_001277079.1 | NP_001264008.1 | 4 - 5 | 433 | 63 | ||
| NM_008088.5 | NP_032114.2 | 4 - 5 | 410 | 63 | ||
| GenBank mRNA | AK017394.1 | - | 4 - 5 | 404 | 63 | |
| AK076926.1 | - | 5 - 6 | 486 | 63 | ||
| AK152149.1 | - | 2 - 3 | 107 | 63 | ||
| AK154463.1 | - | 4 - 5 | 407 | 63 | ||
| BC120528.1 | - | 4 - 5 | 331 | 40129451 | 63 | |
| BC120530.1 | - | 4 - 5 | 331 | 40129453 | 63 | |
| BU053145.1 | - | 4 - 5 | 414 | 63 | ||
| U19860.1 | - | 4 - 5 | 436 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gas7
Genomic Map