Assay Details
| Gene Symbol: Gas7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109657.3 | NP_001103127.1 | 6 - 7 | 746 | 59 | |
| NM_001277079.1 | NP_001264008.1 | 6 - 7 | 575 | 59 | ||
| NM_001277080.1 | NP_001264009.1 | 5 - 6 | 489 | 59 | ||
| NM_008088.5 | NP_032114.2 | 6 - 7 | 552 | 59 | ||
| GenBank mRNA | AF133184.1 | - | 2 - 3 | 424 | 59 | |
| AK017394.1 | - | 6 - 7 | 546 | 59 | ||
| AK076926.1 | - | 7 - 8 | 628 | 59 | ||
| AK152149.1 | - | 4 - 5 | 249 | 59 | ||
| AK154463.1 | - | 6 - 7 | 549 | 59 | ||
| AK172943.1 | - | 2 - 3 | 368 | 59 | ||
| BC120528.1 | - | 6 - 7 | 473 | 40129451 | 59 | |
| BC120530.1 | - | 6 - 7 | 473 | 40129453 | 59 | |
| BU053145.1 | - | 6 - 7 | 556 | 59 | ||
| U19860.1 | - | 6 - 7 | 578 | 59 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gas7
Genomic Map