Assay Details
| Gene Symbol: Gas7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109657.3 | NP_001103127.1 | 9 - 10 | 1019 | 121 | |
| NM_001277079.1 | NP_001264008.1 | 9 - 10 | 848 | 121 | ||
| NM_001277080.1 | NP_001264009.1 | 8 - 9 | 762 | 121 | ||
| NM_008088.5 | NP_032114.2 | 9 - 10 | 825 | 121 | ||
| GenBank mRNA | AF133184.1 | - | 5 - 6 | 697 | 121 | |
| AK017394.1 | - | 9 - 10 | 818 | 121 | ||
| AK076926.1 | - | 10 - 11 | 901 | 121 | ||
| AK152149.1 | - | 7 - 8 | 522 | 121 | ||
| AK154463.1 | - | 9 - 10 | 822 | 121 | ||
| AK172943.1 | - | 5 - 6 | 641 | 121 | ||
| AK200726.1 | - | 2 - 3 | 165 | 121 | ||
| BC120528.1 | - | 9 - 10 | 746 | 40129451 | 121 | |
| BC120530.1 | - | 9 - 10 | 746 | 40129453 | 121 | |
| CV562268.1 | - | 1 - 2 | 63 | 121 | ||
| U19860.1 | - | 9 - 10 | 851 | 121 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gas7
Genomic Map