Assay Details
| Gene Symbol: Usp43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291049.1 | NP_001277978.1 | 11 - 12 | 1766 | 74 | |
| NM_173754.4 | NP_776115.2 | 11 - 12 | 1766 | 74 | ||
| GenBank mRNA | AK042217.1 | - | 11 - 12 | 1767 | 74 | |
| AK083271.1 | - | 11 - 12 | 1604 | 74 | ||
| BC021474.1 | - | 10 - 11 | 1056 | 5343429 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Usp43
Genomic Map