Assay Details
| Gene Symbol: Exd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172857.2 | NP_766445.1 | 10 - 11 | 1485 | 92 | |
| GenBank mRNA | AK030132.1 | - | 10 - 11 | 1487 | 92 | |
| BC092241.1 | - | 8 - 9 | 739 | 30670567 | 92 | |
More Information
Set Membership: |
|

Exd1
Genomic Map