Assay Details
| Gene Symbol: Exd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172857.2 | NP_766445.1 | 11 - 12 | 1626 | 95 | |
| GenBank mRNA | AK030132.1 | - | 11 - 12 | 1628 | 95 | |
| BC092241.1 | - | 9 - 10 | 880 | 30670567 | 95 | |
| BC144750.1 | - | 10 - 11 | 1009 | 9053298 | 95 | |
| BC150662.1 | - | 11 - 12 | 1103 | 9087573 | 95 | |
More Information
Updated Mapping Information
Set Membership: |
|

Exd1
Genomic Map