Assay Details
| Gene Symbol: Exd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172857.2 | NP_766445.1 | 2 - 3 | 721 | 142 | |
| GenBank mRNA | AK030132.1 | - | 2 - 3 | 723 | 142 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK030132.1, NM_172857.2 transcripts.
Set Membership: |
|

Exd1
Genomic Map