Assay Details
| Gene Symbol: Exd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172857.2 | NP_766445.1 | 6 - 7 | 1153 | 77 | |
| GenBank mRNA | AK030132.1 | - | 6 - 7 | 1155 | 77 | |
| BC092241.1 | - | 4 - 5 | 407 | 30670567 | 77 | |
| BC144750.1 | - | 5 - 6 | 536 | 9053298 | 77 | |
| BC150662.1 | - | 6 - 7 | 630 | 9087573 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Exd1
Genomic Map