Assay Details
| Gene Symbol: Exd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172857.2 | NP_766445.1 | 8 - 9 | 1302 | 90 | |
| GenBank mRNA | AK030132.1 | - | 8 - 9 | 1304 | 90 | |
| BC092241.1 | - | 6 - 7 | 556 | 30670567 | 90 | |
| BC144750.1 | - | 7 - 8 | 685 | 9053298 | 90 | |
| BC150662.1 | - | 8 - 9 | 779 | 9087573 | 90 | |
More Information
Updated Mapping Information
Set Membership: |
|

Exd1
Genomic Map