Assay Details
Gene Symbol: Rpap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163701.1 | NP_001157173.1 | 7 - 8 | 1175 | 69 | |
NM_177294.5 | NP_796268.3 | 8 - 9 | 1252 | 69 | ||
GenBank mRNA | AK122505.1 | - | 8 - 9 | 1219 | 69 | |
AK154007.1 | - | 7 - 8 | 1158 | 69 | ||
AK161487.1 | - | 8 - 9 | 1235 | 69 | ||
AK164658.1 | - | 8 - 9 | 1235 | 69 | ||
AK166232.1 | - | 7 - 8 | 1158 | 69 | ||
AK167589.1 | - | 7 - 8 | 1158 | 69 | ||
AK171233.1 | - | 7 - 8 | 1131 | 69 | ||
BC088899.1 | - | 6 - 7 | 980 | 69 | ||
BC117973.1 | - | 7 - 8 | 1144 | 40098643 | 69 | |
BC117974.1 | - | 7 - 8 | 1144 | 40098644 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
