Assay Details
| Gene Symbol: Gorasp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027352.4 | NP_081628.3 | 7 - 8 | 1016 | 58 | |
| NR_027343.1 | 8 - 9 | 1213 | 58 | |||
| GenBank mRNA | AK002515.1 | - | 8 - 9 | 1094 | 58 | |
| AK010664.2 | - | 7 - 8 | 935 | 58 | ||
| AK033413.1 | - | 7 - 8 | 950 | 58 | ||
| AK044691.1 | - | 901 | 58 | |||
| AK077153.1 | - | 7 - 8 | 891 | 58 | ||
| AK077683.1 | - | 7 - 8 | 951 | 58 | ||
| AK077706.1 | - | 7 - 8 | 883 | 58 | ||
| AK088239.1 | - | 7 - 8 | 897 | 58 | ||
| AK148337.1 | - | 7 - 8 | 950 | 58 | ||
| AK151843.1 | - | 7 - 8 | 898 | 58 | ||
| AK152475.1 | - | 7 - 8 | 898 | 58 | ||
| AK152544.1 | - | 7 - 8 | 898 | 58 | ||
| AK152603.1 | - | 7 - 8 | 926 | 58 | ||
| AK153049.1 | - | 7 - 8 | 898 | 58 | ||
| AK153555.1 | - | 7 - 8 | 949 | 58 | ||
| AK159469.1 | - | 7 - 8 | 948 | 58 | ||
| AK167823.1 | - | 7 - 8 | 950 | 58 | ||
| AK168928.1 | - | 8 - 9 | 1146 | 58 | ||
| AK169382.1 | - | 7 - 8 | 898 | 58 | ||
| AK184024.1 | - | 2 - 3 | 273 | 58 | ||
| AK207289.1 | - | 1 - 2 | 111 | 58 | ||
| BC005600.1 | - | 7 - 8 | 889 | 3482776 | 58 | |
| BC016455.1 | - | 6 - 7 | 741 | 4234343 | 58 | |
| BC024982.1 | - | 7 - 8 | 902 | 58 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gorasp2
Genomic Map