Assay Details
| Gene Symbol: Ubr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC172146.1 | - | 13 - 14 | 1757 | 113 | |
| DQ924536.1 | - | 13 - 14 | 2068 | 113 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: Ubr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC172146.1 | - | 13 - 14 | 1757 | 113 | |
| DQ924536.1 | - | 13 - 14 | 2068 | 113 | ||
Set Membership: |
|