Assay Details
| Gene Symbol: Tlk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172664.3 | NP_766252.2 | 11 - 12 | 1316 | 126 | |
| GenBank mRNA | AK029773.1 | - | 10 - 11 | 1042 | 126 | |
| AK172897.1 | - | 5 - 6 | 580 | 126 | ||
| BC033931.1 | - | 9 - 10 | 878 | 126 | ||
| BC051641.1 | - | 11 - 12 | 1316 | 6507968 | 126 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tlk1
Genomic Map