Assay Details
| Gene Symbol: Tlk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172664.3 | NP_766252.2 | 3 - 4 | 475 | 122 | |
| GenBank mRNA | AK029773.1 | - | 2 - 3 | 201 | 122 | |
| BC051641.1 | - | 3 - 4 | 475 | 6507968 | 122 | |
More Information
Set Membership: |
|

Tlk1
Genomic Map