Assay Details
| Gene Symbol: Tlk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172664.3 | NP_766252.2 | 7 - 8 | 793 | 113 | |
| GenBank mRNA | AK029773.1 | - | 6 - 7 | 519 | 113 | |
| AK172897.1 | - | 1 - 2 | 57 | 113 | ||
| BC033931.1 | - | 5 - 6 | 355 | 113 | ||
| BC051641.1 | - | 7 - 8 | 793 | 6507968 | 113 | |
| BC057369.1 | - | 8 - 9 | 779 | 113 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tlk1
Genomic Map