Assay Details
| Gene Symbol: Tubgcp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146190.2 | NP_666302.2 | 8 - 9 | 875 | 103 | |
| GenBank mRNA | AK032031.1 | - | 8 - 9 | 835 | 103 | |
| AK044444.1 | - | 8 - 9 | 761 | 103 | ||
| AK044887.1 | - | 8 - 9 | 859 | 103 | ||
| AK051326.1 | - | 8 - 9 | 870 | 103 | ||
| AK086158.1 | - | 8 - 9 | 834 | 104 | ||
| AK151454.1 | - | 8 - 9 | 833 | 103 | ||
| AK152033.1 | - | 8 - 9 | 833 | 103 | ||
| AK153331.1 | - | 8 - 9 | 833 | 103 | ||
| AK153357.1 | - | 8 - 9 | 833 | 103 | ||
| AK160939.1 | - | 8 - 9 | 789 | 103 | ||
| AK161681.1 | - | 8 - 9 | 833 | 103 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK086158.1 transcripts.
Set Membership: |
|

Tubgcp5
Genomic Map