Assay Details
| Gene Symbol: Galnt3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015736.2 | NP_056551.2 | 1 - 2 | 272 | 123 | |
| GenBank mRNA | AK007135.1 | - | 1 - 2 | 238 | 123 | |
| AK019995.1 | - | 1 - 2 | 272 | 123 | ||
| AK135489.1 | - | 1 - 2 | 233 | 123 | ||
| BC043331.1 | - | 1 - 2 | 128 | 5342768 | 123 | |
| U70538.1 | - | 1 - 2 | 116 | 123 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK135489.1, BC043331.1, NM_015736.2, U70538.1 transcripts.
Set Membership: |
|

Galnt3
Genomic Map