Assay Details
| Gene Symbol: Sco1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040026.1 | NP_001035115.1 | 3 - 4 | 529 | 88 | |
| GenBank mRNA | AK011251.1 | - | 3 - 4 | 545 | 88 | |
| AK017794.1 | - | 3 - 4 | 530 | 88 | ||
| AK139512.1 | - | 3 - 4 | 500 | 88 | ||
| AK167305.1 | - | 3 - 4 | 394 | 88 | ||
| BC139008.1 | - | 3 - 4 | 524 | 8862030 | 88 | |
| BC139009.1 | - | 3 - 4 | 524 | 8862031 | 88 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sco1
Genomic Map