Assay Details
| Gene Symbol: Kcnh7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133207.2 | NP_573470.2 | 12 - 13 | 2904 | 68 | |
| GenBank mRNA | AJ291608.1 | - | 12 - 13 | 2711 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Kcnh7
Genomic Map