| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0200031034.mm.1 |
NM_133207.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200031034.mm.1 |
NM_133207.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200031034.mm.1 |
TC0200003632.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200031033.mm.1 |
NM_133207.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200031033.mm.1 |
NM_133207.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200031033.mm.1 |
TC0200003632.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200031037.mm.1 |
NM_133207.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200031037.mm.1 |
NM_133207.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200031037.mm.1 |
TC0200003632.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200031036.mm.1 |
NM_133207.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200031036.mm.1 |
NM_133207.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200031036.mm.1 |
TC0200003632.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200031038.mm.1 |
NM_133207.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200031038.mm.1 |
NM_133207.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200031038.mm.1 |
TC0200003632.mm.1 |
MTA TC |