Assay Details
| Gene Symbol: Kcnh7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133207.2 | NP_573470.2 | 9 - 10 | 2363 | 85 | |
| GenBank mRNA | AJ291608.1 | - | 9 - 10 | 2170 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Kcnh7
Genomic Map