Assay Details
| Gene Symbol: Mastl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025979.4 | NP_080255.3 | 6 - 7 | 967 | 121 | |
| GenBank mRNA | AK006448.1 | - | 1 - 2 | 68 | 121 | |
| AK012597.1 | - | 1 - 2 | 68 | 121 | ||
| AK030140.1 | - | 6 - 7 | 862 | 121 | ||
| AK078365.1 | - | 6 - 7 | 841 | 121 | ||
| BC066834.1 | - | 224 | 121 | |||
| BC086483.1 | - | 236 | 121 | |||
| BC103779.1 | - | 6 - 7 | 843 | 30480071 | 121 | |
| CD546688.1 | - | 2 - 3 | 190 | 121 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mastl
Genomic Map