Assay Details
| Gene Symbol: Abi1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077192.2 | NP_001070660.1 | 3 - 4 | 645 | 63 | |
| NM_001077193.2 | NP_001070661.1 | 3 - 4 | 645 | 63 | ||
| NM_007380.3 | NP_031406.2 | 3 - 4 | 645 | 63 | ||
| NM_145994.2 | NP_666106.1 | 3 - 4 | 645 | 63 | ||
| GenBank mRNA | AF420251.1 | - | 3 - 4 | 581 | 63 | |
| AK151026.1 | - | 3 - 4 | 553 | 63 | ||
| AK151363.1 | - | 3 - 4 | 624 | 63 | ||
| AK152061.1 | - | 3 - 4 | 509 | 63 | ||
| AK152184.1 | - | 3 - 4 | 509 | 63 | ||
| AK153130.1 | - | 3 - 4 | 553 | 63 | ||
| AK164040.1 | - | 3 - 4 | 561 | 63 | ||
| AK167328.1 | - | 3 - 4 | 604 | 63 | ||
| AK167568.1 | - | 3 - 4 | 600 | 63 | ||
| AK167744.1 | - | 3 - 4 | 592 | 63 | ||
| AK184875.1 | - | 1 - 2 | 89 | 63 | ||
| AK190191.1 | - | 1 - 2 | 89 | 63 | ||
| AY033645.1 | - | 3 - 4 | 489 | 63 | ||
| BC004657.1 | - | 3 - 4 | 596 | 3498068 | 63 | |
| BC079642.1 | - | 3 - 4 | 551 | 6816252 | 63 | |
| U17698.1 | - | 2 - 3 | 271 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Abi1
Genomic Map