Assay Details
| Gene Symbol: Abi1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077190.2 | NP_001070658.1 | 5 - 6 | 753 | 91 | |
| NM_001077192.2 | NP_001070660.1 | 4 - 5 | 738 | 91 | ||
| NM_001077193.2 | NP_001070661.1 | 4 - 5 | 738 | 91 | ||
| NM_007380.3 | NP_031406.2 | 4 - 5 | 738 | 91 | ||
| NM_145994.2 | NP_666106.1 | 4 - 5 | 738 | 91 | ||
| GenBank mRNA | AF420251.1 | - | 4 - 5 | 674 | 91 | |
| AK034476.1 | - | 5 - 6 | 635 | 91 | ||
| AK151026.1 | - | 4 - 5 | 646 | 91 | ||
| AK151363.1 | - | 4 - 5 | 717 | 91 | ||
| AK152046.1 | - | 3 - 4 | 673 | 91 | ||
| AK152061.1 | - | 4 - 5 | 602 | 91 | ||
| AK152184.1 | - | 4 - 5 | 602 | 91 | ||
| AK153130.1 | - | 4 - 5 | 646 | 91 | ||
| AK164040.1 | - | 4 - 5 | 654 | 91 | ||
| AK165924.1 | - | 4 - 5 | 711 | 91 | ||
| AK167328.1 | - | 4 - 5 | 697 | 91 | ||
| AK167568.1 | - | 4 - 5 | 693 | 91 | ||
| AK167744.1 | - | 4 - 5 | 685 | 91 | ||
| AK181529.1 | - | 3 - 4 | 281 | 91 | ||
| AK184875.1 | - | 2 - 3 | 182 | 91 | ||
| AK201770.1 | - | 3 - 4 | 197 | 91 | ||
| AK215001.1 | - | 3 - 4 | 197 | 91 | ||
| AY033645.1 | - | 4 - 5 | 582 | 91 | ||
| BC004657.1 | - | 4 - 5 | 689 | 3498068 | 91 | |
| BC079642.1 | - | 4 - 5 | 644 | 6816252 | 91 | |
| U17698.1 | - | 3 - 4 | 364 | 91 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Abi1
Genomic Map