Assay Details
| Gene Symbol: Abi1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077190.2 | NP_001070658.1 | 8 - 9 | 1173 | 101 | |
| NM_007380.3 | NP_031406.2 | 7 - 8 | 1155 | 101 | ||
| NM_145994.2 | NP_666106.1 | 7 - 8 | 1158 | 101 | ||
| GenBank mRNA | AF420251.1 | - | 7 - 8 | 1091 | 101 | |
| AK034476.1 | - | 8 - 9 | 1055 | 101 | ||
| AK167328.1 | - | 7 - 8 | 1114 | 101 | ||
| AK167568.1 | - | 7 - 8 | 1113 | 101 | ||
| AK179494.1 | - | 1 - 2 | 59 | 101 | ||
| AK182193.1 | - | 1 - 2 | 59 | 101 | ||
| BC004657.1 | - | 7 - 8 | 1109 | 3498068 | 101 | |
More Information
Updated Mapping Information
Set Membership: |
|

Abi1
Genomic Map