Assay Details
| Gene Symbol: Gad2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008078.2 | NP_032104.2 | 14 - 15 | 1829 | 69 | |
| GenBank mRNA | AF326549.1 | - | 14 - 15 | 1493 | 69 | |
| AF326550.1 | - | 14 - 15 | 1493 | 69 | ||
| BC018380.1 | - | 14 - 15 | 1544 | 4482097 | 69 | |
| KC134211.1 | - | 14 - 15 | 1493 | 69 | ||
| L16980.1 | - | 14 - 15 | 1565 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gad2
Genomic Map