Assay Details
| Gene Symbol: Gad2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008078.2 | NP_032104.2 | 2 - 3 | 471 | 53 | |
| GenBank mRNA | AF326549.1 | - | 2 - 3 | 135 | 53 | |
| AF326550.1 | - | 2 - 3 | 135 | 53 | ||
| AK136834.1 | - | 2 - 3 | 171 | 53 | ||
| BC018380.1 | - | 2 - 3 | 186 | 4482097 | 53 | |
| BQ571205.1 | - | 2 - 3 | 579 | 53 | ||
| D42051.1 | - | 2 - 3 | 487 | 53 | ||
| KC134211.1 | - | 2 - 3 | 135 | 53 | ||
| L16980.1 | - | 2 - 3 | 207 | 53 | ||
More Information
Set Membership: |
|

Gad2
Genomic Map