| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0200001780.mm.1 |
NM_001102436.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200001780.mm.1 |
NM_001102436.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200001780.mm.1 |
NM_001102437.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200001780.mm.1 |
NM_001102437.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200001780.mm.1 |
NM_001102438.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200001780.mm.1 |
NM_001102438.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200001780.mm.1 |
NM_028793.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200001780.mm.1 |
NM_028793.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200001780.mm.1 |
TC0200000361.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200001781.mm.1 |
NM_001102436.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200001781.mm.1 |
NM_001102436.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200001781.mm.1 |
NM_001102437.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200001781.mm.1 |
NM_001102437.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200001781.mm.1 |
NM_001102438.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200001781.mm.1 |
NM_001102438.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200001781.mm.1 |
NM_028793.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200001781.mm.1 |
NM_028793.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200001781.mm.1 |
TC0200000361.mm.1 |
MTA TC |