Assay Details
| Gene Symbol: Mrc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008625.2 | NP_032651.2 | 28 - 29 | 4177 | 115 | |
| GenBank mRNA | AK079897.1 | - | 1 - 2 | 129 | 115 | |
| BC141338.1 | - | 28 - 29 | 4129 | 9055950 | 115 | |
| Z11974.1 | - | 28 - 29 | 4177 | 115 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mrc1
Genomic Map