Assay Details
| Gene Symbol: Cds2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291039.1 | NP_001277968.1 | 11 - 12 | 1071 | 63 | |
| NM_138651.7 | NP_619592.1 | 11 - 12 | 1394 | 63 | ||
| GenBank mRNA | AK036328.1 | - | 11 - 12 | 1245 | 63 | |
| AK049257.1 | - | 11 - 12 | 1072 | 63 | ||
| AK147541.1 | - | 11 - 12 | 1286 | 63 | ||
| AK166001.1 | - | 11 - 12 | 1245 | 63 | ||
| AY159802.1 | - | 11 - 12 | 1317 | 63 | ||
| BC003852.1 | - | 11 - 12 | 1300 | 3600825 | 63 | |
| BC069879.1 | - | 11 - 12 | 1273 | 30439069 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cds2
Genomic Map