Assay Details
| Gene Symbol: Cds2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291040.1 | NP_001277969.1 | 1 - 2 | 351 | 84 | |
| NM_138651.7 | NP_619592.1 | 1 - 2 | 351 | 84 | ||
| GenBank mRNA | AK036328.1 | - | 1 - 2 | 202 | 84 | |
| AK050589.1 | - | 1 - 2 | 259 | 84 | ||
| AK147541.1 | - | 1 - 2 | 243 | 84 | ||
| AK166001.1 | - | 1 - 2 | 202 | 84 | ||
| AK170888.1 | - | 1 - 2 | 240 | 84 | ||
| AY159802.1 | - | 1 - 2 | 274 | 84 | ||
| BC003852.1 | - | 1 - 2 | 257 | 3600825 | 84 | |
| BC059776.1 | - | 1 - 2 | 154 | 84 | ||
| BC069879.1 | - | 1 - 2 | 230 | 30439069 | 84 | |
| CJ112406.1 | - | 1 - 2 | 352 | 84 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cds2
Genomic Map