Assay Details
| Gene Symbol: Yme1l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013771.5 | NP_038799.1 | 14 - 15 | 1757 | 139 | |
| GenBank mRNA | AF090430.1 | - | 14 - 15 | 1735 | 139 | |
| AK079175.1 | - | 14 - 15 | 1717 | 139 | ||
| AY136286.1 | - | 14 - 15 | 1573 | 139 | ||
| AY136287.1 | - | 14 - 15 | 1726 | 139 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Yme1l1
Genomic Map