Assay Details
| Gene Symbol: Yme1l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013771.5 | NP_038799.1 | 16 - 17 | 2030 | 123 | |
| GenBank mRNA | AF090430.1 | - | 16 - 17 | 2008 | 123 | |
| AK045141.1 | - | 16 - 17 | 2014 | 123 | ||
| AK079175.1 | - | 16 - 17 | 1990 | 123 | ||
| AY136286.1 | - | 16 - 17 | 1846 | 123 | ||
| AY136287.1 | - | 16 - 17 | 1999 | 123 | ||
| BC007128.1 | - | 16 - 17 | 1991 | 3591123 | 123 | |
More Information
Updated Mapping Information
Set Membership: |
|

Yme1l1
Genomic Map