Assay Details
| Gene Symbol: Yme1l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013771.5 | NP_038799.1 | 3 - 4 | 519 | 88 | |
| GenBank mRNA | AF090430.1 | - | 3 - 4 | 497 | 88 | |
| AK032273.1 | - | 3 - 4 | 507 | 88 | ||
| AK045141.1 | - | 3 - 4 | 504 | 88 | ||
| AK079175.1 | - | 3 - 4 | 479 | 88 | ||
| AK137665.1 | - | 3 - 4 | 471 | 88 | ||
| AY136286.1 | - | 3 - 4 | 335 | 88 | ||
| AY136287.1 | - | 3 - 4 | 488 | 88 | ||
| BC007128.1 | - | 3 - 4 | 480 | 3591123 | 88 | |
More Information
Updated Mapping Information
Set Membership: |
|

Yme1l1
Genomic Map